(1R)-1-(4-Chlorophenyl)-1,2-ethanediol 95%
Lieferant: Apollo Scientific
OR947779-1G
APOSOR947779-1GEA
574
EUR
APOSOR947779-1G
(1R)-1-(4-Chlorophenyl)-1,2-ethanediol 95%
(1R)-1-(4-Chlorophenyl)-1,2-ethanediol
Formel:
C₈H₉ClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD19689566 CAS-Nummer: 152142-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...