Ac-Ile-NHMe 95%

Lieferant: Apollo Scientific

OR947952-1G
APOSOR947952-1GEA 243 EUR
APOSOR947952-1G
Ac-Ile-NHMe 95%
Ac-Ile-NHMe
Formel: C₉H₁₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00237495
CAS-Nummer: 32483-16-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR