(S)-Methyl 2-amino-3-chloropropanoate hydrochloride 95%

Lieferant: Apollo Scientific

OR947974-5G
APOSOR947974-5GEA 0 EUR
APOSOR947974-5G
(S)-Methyl 2-amino-3-chloropropanoate hydrochloride 95%
(S)-Methyl 2-amino-3-chloropropanoate hydrochloride
Formel: C₄H₈ClNO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD00066119
CAS-Nummer: 112346-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR