2-Chloro-4-methoxypyridin-3-amine 95%
Lieferant: Apollo Scientific
OR948057-1G
APOSOR948057-1GEA
632
EUR
APOSOR948057-1G
2-Chloro-4-methoxypyridin-3-amine 95%
2-Chloro-4-methoxypyridin-3-amine
Formel:
C₆H₇ClN₂O Molecular Weight: 158,586 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699223 CAS-Nummer: 173435-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...