(3-Chlorpropyl)(dimethoxy)methylsilan 95%

Lieferant: Apollo Scientific

Synonyme: 3-(Methyldimethoxysilyl)propyl Chloride, (3-Chloropropyl)dimethoxy(methyl)silane

OR948176-100G
APOSOR948176-100GEA 63.8 EUR
APOSOR948176-100G
(3-Chlorpropyl)(dimethoxy)methylsilan 95%
(3-Chlorpropyl)(dimethoxy)methylsilan
Formel: C₆H₁₅ClO₂Si
Molecular Weight: 182.72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00053912
CAS-Nummer: 18171-19-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR