Chloromethyl(dimethyl)methoxysilane 95%

Lieferant: Apollo Scientific

OR948192-1G
APOSOR948192-1GEA 647 EUR
APOSOR948192-1G
Chloromethyl(dimethyl)methoxysilane 95%
Chloromethyl(dimethyl)methoxysilane
Formel: C₄H₁₁ClOSi
Molecular Weight: 138,668 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664349
CAS-Nummer: 18143-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR