2,6-Dichloro-N-methylpyridin-4-amine 95%

Lieferant: Apollo Scientific

OR948201-1G
APOSOR948201-1GEA 108 EUR
APOSOR948201-1G
2,6-Dichloro-N-methylpyridin-4-amine 95%
2,6-Dichloro-N-methylpyridin-4-amine
Formel: C₆H₆Cl₂N₂
Molecular Weight: 177,031 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12024871
CAS-Nummer: 175461-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR