N-Formyl-L-leucine 95%

Lieferant: Apollo Scientific

Synonyme: N-Formyl-Leu-OH, For-Leu-OH

OR948218-1G
APOSOR948218-1GEA 50.9 EUR
APOSOR948218-1G
N-Formyl-L-leucine 95%
N-Formyl-L-leucine
Formel: C₇H₁₃NO₃
Molecular Weight: 159,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00055861
CAS-Nummer: 6113-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR