8-Methoxychroman-4-one 95%

Lieferant: Apollo Scientific

OR948291-1G
APOSOR948291-1GEA 108 EUR
APOSOR948291-1G
8-Methoxychroman-4-one 95%
8-Methoxychroman-4-one
Formel: C₁₀H₁₀O₃
Molecular Weight: 178.185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00094977
CAS-Nummer: 20351-79-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR