4,6-Dimethyl-2,3-dihydropyridazin-3-one

Lieferant: Apollo Scientific

OR948323-1G OR948323-250MG OR948323-500MG
APOSOR948323-1GEA 899 EUR
APOSOR948323-1G APOSOR948323-250MG APOSOR948323-500MG
4,6-Dimethyl-2,3-dihydropyridazin-3-one
4,6-Dimethyl-2,3-dihydropyridazin-3-one
Formel: C₆H₈N₂O
Molecular Weight: 124.141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00867544
CAS-Nummer: 7007-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR