Boc-(R)-2-Cyclobutylglycine 97%
Lieferant: Apollo Scientific
OR948353-1G
OR948353-250MG
OR948353-500MG
APOSOR948353-1GEA
810
EUR
APOSOR948353-1G
APOSOR948353-250MG
APOSOR948353-500MG
Boc-(R)-2-Cyclobutylglycine 97%
Boc-(R)-2-Cyclobutylglycine
Formel:
C₁₁H₁₉NO₄ Molecular Weight: 229.273 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659974 CAS-Nummer: 155905-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...