2-(Benzyloxy)cyclobutan-1-one

Lieferant: Apollo Scientific

Synonyme: 2-(Benzyloxy)cyclobutanone

OR948453-1G
APOSOR948453-1GEA 532 EUR
APOSOR948453-1G
2-(Benzyloxy)cyclobutan-1-one
2-(Benzyloxy)cyclobutan-1-one
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09908029
CAS-Nummer: 206751-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR