1-[4-(Ethylsulphonyl)phenyl]methanamine
Lieferant: Apollo Scientific
|
Achtung
|
OR948469-250MG
APOSOR948469-250MGEA
21.9
EUR
APOSOR948469-250MG
1-[4-(Ethylsulphonyl)phenyl]methanamine
1-[4-(Ethylsulphonyl)phenyl]methanamine
Formel:
C₉H₁₃NO₂S Molecular Weight: 199,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10032480 CAS-Nummer: 583837-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...