3-(Chloromethyl)cyclobutan-1-one

Lieferant: Apollo Scientific

OR948551-1G
APOSOR948551-1GEA 130 EUR
APOSOR948551-1G
3-(Chloromethyl)cyclobutan-1-one
3-(Chloromethyl)cyclobutan-1-one
Formel: C₅H₇ClO
Molecular Weight: 118,561 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858225
CAS-Nummer: 175021-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR