1-(2-Methoxyethyl)-1H-pyrazole-4-carbaldehyde 95%

Lieferant: Apollo Scientific

OR948690-1G
APOSOR948690-1GEA 306 EUR
APOSOR948690-1G
1-(2-Methoxyethyl)-1H-pyrazole-4-carbaldehyde 95%
1-(2-Methoxyethyl)-1H-pyrazole-4-carbaldehyde
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13248693
CAS-Nummer: 304693-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR