2-Methylimidazo[1,2-a]pyridin-3-carbonsäure

Lieferant: Apollo Scientific
Achtung

OR9488-250MG
APOSOR9488-250MGEA 24.8 EUR
APOSOR9488-250MG
2-Methylimidazo[1,2-a]pyridin-3-carbonsäure
2-Methylimidazo[1,2-a]pyridin-3-carbonsäure
Formel: C₉H₈N₂O₂
Molecular Weight: 176,1700 g/mol
Schmelzpunkt: 147…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00244615
CAS-Nummer: 21801-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR