3-(Bromomethyl)-1,1-dimethylcyclobutane

Lieferant: Apollo Scientific

OR948832-1G
APOSOR948832-1GEA 516 EUR
APOSOR948832-1G
3-(Bromomethyl)-1,1-dimethylcyclobutane
3-(Bromomethyl)-1,1-dimethylcyclobutane
Formel: C₇H₁₃Br
Lagertemperatur: Raumtemperatur
MDL: MFCD17170399
CAS-Nummer: 76207-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR