5-Methyl-3-pyridinboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methylpyridine-3-boronic acid

OR9489-1G OR9489-5G OR9489-10G
APOSOR9489-1GEA 21.9 EUR
APOSOR9489-1G APOSOR9489-5G APOSOR9489-10G
5-Methyl-3-pyridinboronsäure 97%
5-Methyl-3-pyridineboronic acid
Formel: C₆H₈BNO₂
Molecular Weight: 136.95 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD03428528
CAS-Nummer: 173999-18-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR