Methyl 2-methylazetidine-2-carboxylate hydrochloride

Lieferant: Apollo Scientific

OR948948-1G
APOSOR948948-1GEA 1270 EUR
APOSOR948948-1G
Methyl 2-methylazetidine-2-carboxylate hydrochloride
Methyl 2-methylazetidine-2-carboxylate hydrochloride
Formel: C₆H₁₁NO₂·ClH
Molecular Weight: 165,618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18812894
CAS-Nummer: 309978-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR