Spiro[2.5]octan-6-ol

Lieferant: Apollo Scientific

OR949206-1G
APOSOR949206-1GEA 1830 EUR
APOSOR949206-1G
Spiro[2.5]octan-6-ol
Spiro[2.5]octan-6-ol
Formel: C₈H₁₄O
Molecular Weight: 126,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22418483
CAS-Nummer: 22428-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR