2-Azaspiro[4.5]decane-7-carboxylic acid

Lieferant: Apollo Scientific

OR949228-1G OR949228-250MG OR949228-500MG
APOSOR949228-1GEA 1110 EUR
APOSOR949228-1G APOSOR949228-250MG APOSOR949228-500MG
2-Azaspiro[4.5]decane-7-carboxylic acid
2-Azaspiro[4.5]decane-7-carboxylic acid
Formel: C₁₀H₁₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22566122
CAS-Nummer: 1363382-89-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR