tert-Butyl (2S)-2-(bromomethyl)azetidine-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: (S)-tert-Butyl 2-(bromomethyl)azetidine-1-carboxylate

OR949254-1G
APOSOR949254-1GEA 772 EUR
APOSOR949254-1G
tert-Butyl (2S)-2-(bromomethyl)azetidine-1-carboxylate
tert-Butyl (2S)-2-(bromomethyl)azetidine-1-carboxylate
Formel: C₉H₁₆BrNO₂
Molecular Weight: 250,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22566238
CAS-Nummer: 1363378-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR