Ethyl rel-(1S,3R)-3-hydroxy-1-methylcyclobutane-1-carboxylate

Lieferant: Apollo Scientific

OR949315-1G
APOSOR949315-1GEA 1490 EUR
APOSOR949315-1G
Ethyl rel-(1S,3R)-3-hydroxy-1-methylcyclobutane-1-carboxylate
Ethyl rel-(1S,3R)-3-hydroxy-1-methylcyclobutane-1-carboxylate
Formel: C₈H₁₄O₃
Molecular Weight: 158,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23105926
CAS-Nummer: 1408074-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR