3-Ethoxycyclobutan-1-amine hydrochloride

Lieferant: Apollo Scientific

OR949359-1G
APOSOR949359-1GEA 811 EUR
APOSOR949359-1G
3-Ethoxycyclobutan-1-amine hydrochloride
3-Ethoxycyclobutan-1-amine hydrochloride
Formel: C₆H₁₃NO·ClH
Molecular Weight: 151,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23106133
CAS-Nummer: 1408076-38-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR