2-Methylbenzo[d]oxazol-6-ol 95%

Lieferant: Apollo Scientific

OR949663-1G OR949663-250MG
APOSOR949663-1GEA 415 EUR
APOSOR949663-1G APOSOR949663-250MG
2-Methylbenzo[d]oxazol-6-ol 95%
2-Methylbenzo[d]oxazol-6-ol
Formel: C₈H₇NO₂
Molecular Weight: 149.147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00464862
CAS-Nummer: 5078-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR