2-(2-Methyl-2-butanyl)phenol 95%

Lieferant: Apollo Scientific

OR949676-25G
APOSOR949676-25GEA 132 EUR
APOSOR949676-25G
2-(2-Methyl-2-butanyl)phenol 95%
2-tert-Amylphenol
Formel: C₁₁H₁₆O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 3279-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR