6-Methoxy-2-methylnicotinaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 6-Methoxy-2-methylpyridine-3-carboxaldehyde

OR949793-1G
APOSOR949793-1GEA 521 EUR
APOSOR949793-1G
6-Methoxy-2-methylnicotinaldehyde 95%
6-Methoxy-2-methylnicotinaldehyde
Formel: C₈H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11044370
CAS-Nummer: 156094-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR