(R)-2-Acetamido-2-phenylacetic acid 95%

Lieferant: Apollo Scientific

OR949797-1G
APOSOR949797-1GEA 72.5 EUR
APOSOR949797-1G
(R)-2-Acetamido-2-phenylacetic acid 95%
(R)-2-Acetamido-2-phenylacetic acid
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12795908
CAS-Nummer: 14257-84-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR