2-Ethoxy-3-pyridinboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Ethoxypyridin-3-yl)boronic acid
OR9499-1G
OR9499-25G
OR9499-5G
APOSOR9499-1GEA
21.9
EUR
APOSOR9499-1G
APOSOR9499-25G
APOSOR9499-5G
2-Ethoxy-3-pyridinboronsäure
2-Ethoxy-3-pyridineboronic acid
Formel:
C₇H₁₀BNO₃ Molecular Weight: 166.97 g/mol Schmelzpunkt: 102…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06201036 CAS-Nummer: 854373-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...