1-Methyl-3-(m-tolyl)-1H-pyrazol-5-amine ≥98%

Lieferant: Apollo Scientific

OR949913-250MG
APOSOR949913-250MGEA 122 EUR
APOSOR949913-250MG
1-Methyl-3-(m-tolyl)-1H-pyrazol-5-amine ≥98%
1-Methyl-3-(m-tolyl)-1H-pyrazol-5-amine
Formel: C₁₁H₁₃N₃
Molecular Weight: 187,241 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06637389
CAS-Nummer: 1017665-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR