3-Formyl-2-methylbenzonitrile 95%

Lieferant: Apollo Scientific

OR949937-1G
APOSOR949937-1GEA 464 EUR
APOSOR949937-1G
3-Formyl-2-methylbenzonitrile 95%
3-Formyl-2-methylbenzonitrile
Formel: C₉H₇NO
Molecular Weight: 145.158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18394296
CAS-Nummer: 27613-34-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR