1-Benzylpyrrolidine-2,5-dione 95%

Lieferant: Apollo Scientific

OR949957-5G
APOSOR949957-5GEA 49.3 EUR
APOSOR949957-5G
1-Benzylpyrrolidine-2,5-dione 95%
1-Benzylpyrrolidine-2,5-dione
Formel: C₁₁H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00516834
CAS-Nummer: 2142-06-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR