N-[(6-Chloropyridin-3-yl)methyl]acetamide 95%

Lieferant: Apollo Scientific

OR949995-1G
APOSOR949995-1GEA 502 EUR
APOSOR949995-1G
N-[(6-Chloropyridin-3-yl)methyl]acetamide 95%
N-[(6-Chloropyridin-3-yl)methyl]acetamide
Formel: C₈H₉ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD13188578
CAS-Nummer: 175424-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR