3-Chloropyridine-4-carboxylic acid ethyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-chloroisonicotinate

OR950143-5G
APOSOR950143-5GEA 92.8 EUR
APOSOR950143-5G
3-Chloropyridine-4-carboxylic acid ethyl ester 95%
3-Chloropyridine-4-carboxylic acid ethyl ester
Formel: C₈H₈ClNO₂
Molecular Weight: 185.608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03411706
CAS-Nummer: 211678-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR