3-Ethyl-3-methylglutarimide 95%

Lieferant: Apollo Scientific

Synonyme: Bemegride, 4-Ethyl-4-methylpiperidine-2,6-dione

OR950148-5G
APOSOR950148-5GEA 213 EUR
APOSOR950148-5G
3-Ethyl-3-methylglutarimide 95%
3-Ethyl-3-methylglutarimide
Formel: C₈H₁₃NO₂
Molecular Weight: 155.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006673
CAS-Nummer: 64-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR