2-[3-(Methylsulphonyl)phenyl]ethylamine 95%

Lieferant: Apollo Scientific
Achtung

OR950154-250MG
APOSOR950154-250MGEA 154 EUR
APOSOR950154-250MG
2-[3-(Methylsulphonyl)phenyl]ethylamine 95%
2-[3-(Methylsulphonyl)phenyl]ethylamine
Formel: C₉H₁₃NO₂S
Molecular Weight: 199,27 g/mol
MDL: MFCD13190314
CAS-Nummer: 933696-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR