4-Amino-3,5-dichlorophenol
Lieferant: Apollo Scientific
|
Achtung
|
OR950194-250MG
APOSOR950194-250MGEA
219
EUR
APOSOR950194-250MG
4-Amino-3,5-dichlorophenol
4-Amino-3,5-dichlorophenol
Formel:
C₆H₅Cl₂NO Molecular Weight: 178,016 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20690586 CAS-Nummer: 26271-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...