2-(Allyloxy)phenol 95%

Lieferant: Apollo Scientific

OR950199-1G
APOSOR950199-1GEA 0 EUR
APOSOR950199-1G
2-(Allyloxy)phenol 95%
2-(Allyloxy)phenol
Formel: C₉H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00072617
CAS-Nummer: 1126-20-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR