4-Cyclopropylbenzenesulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

OR950305-1G OR950305-250MG OR950305-5G
APOSOR950305-1GEA 0 EUR
APOSOR950305-1G APOSOR950305-250MG APOSOR950305-5G
4-Cyclopropylbenzenesulphonyl chloride 95%
4-Cyclopropylbenzenesulphonyl chloride
Formel: C₉H₉ClO₂S
Lagertemperatur: Kühlschrank
MDL: MFCD09929602
CAS-Nummer: 167404-32-2
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR