2-Chloro-3,3-diethoxyprop-1-ene 95%

Lieferant: Apollo Scientific

OR950581-1G
APOSOR950581-1GEA 719 EUR
APOSOR950581-1G
2-Chloro-3,3-diethoxyprop-1-ene 95%
2-Chloro-3,3-diethoxyprop-1-ene
Formel: C₇H₁₃ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23102091
CAS-Nummer: 7575-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR