2-(Chloromethyl)-5-methoxypyridine 95%

Lieferant: Apollo Scientific

OR950620-1G
APOSOR950620-1GEA 502 EUR
APOSOR950620-1G
2-(Chloromethyl)-5-methoxypyridine 95%
2-(Chloromethyl)-5-methoxypyridine
Formel: C₇H₈ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD10697536
CAS-Nummer: 75342-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR