6-Chloro-3,4-dihydro-1(2H)-pyridinecarbonyl chloride 95%
Lieferant: Apollo Scientific
OR950622-1G
APOSOR950622-1GEA
574
EUR
APOSOR950622-1G
6-Chloro-3,4-dihydro-1(2H)-pyridinecarbonyl chloride 95%
6-Chloro-3,4-dihydro-1(2H)-pyridinecarbonyl chloride
Formel:
C₆H₇Cl₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD22575235 CAS-Nummer: 5100-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...