3,5-Dimethoxypicolinonitrile 95%

Lieferant: Apollo Scientific

OR950635-1G
APOSOR950635-1GEA 169 EUR
APOSOR950635-1G
3,5-Dimethoxypicolinonitrile 95%
3,5-Dimethoxypicolinonitrile
Formel: C₈H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18255817
CAS-Nummer: 36057-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR