5-Bromo-N-methylpyridin-3-amine 95%

Lieferant: Apollo Scientific

OR950661-1G
APOSOR950661-1GEA 140 EUR
APOSOR950661-1G
5-Bromo-N-methylpyridin-3-amine 95%
5-Bromo-N-methylpyridin-3-amine
Formel: C₆H₇BrN₂
Molecular Weight: 187,037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13190575
CAS-Nummer: 873383-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR