2-Amino-3-(m-tolyl)propanoic acid 97%

Lieferant: Apollo Scientific

Synonyme: 3-Methyl-DL-phenylalanine

OR950694-1G OR950694-25G OR950694-5G
APOSOR950694-1GEA 26.1 EUR
APOSOR950694-1G APOSOR950694-25G APOSOR950694-5G
2-Amino-3-(m-tolyl)propanoic acid 97%
2-Amino-3-(m-tolyl)propanoic acid
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05227937
CAS-Nummer: 5472-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR