[1,2,4]triazolo[1,5-a]pyrazin-2-amine 97%

Lieferant: Apollo Scientific

Synonyme: [1,2,4]Triazolo[1,5-a]pyrazin-2-amine

OR950865-1G OR950865-250MG
APOSOR950865-1GEA 49.7 EUR
APOSOR950865-1G APOSOR950865-250MG
[1,2,4]triazolo[1,5-a]pyrazin-2-amine 97%
[1,2,4]triazolo[1,5-a]pyrazin-2-amine
Formel: C₅H₅N₅
Molecular Weight: 135.127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847742
CAS-Nummer: 88002-33-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR