DL-2-Hydroxybutansäure 95%

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxybutyric acid, 2-Hydroxybutanoic acid

OR950886-1G
APOSOR950886-1GEA 85.7 EUR
APOSOR950886-1G
DL-2-Hydroxybutansäure 95%
DL-2-Hydroxybutyric acid
Formel: C₄H₈O₃
Molecular Weight: 104,11 g/mol
Siedepunkt: 260 °C
Schmelzpunkt: 43 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00070502
CAS-Nummer: 600-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR