4-Chlor-1,3-dimethyl-1H-pyrazol 95%

Lieferant: Apollo Scientific

OR950948-1G
APOSOR950948-1GEA 85.7 EUR
APOSOR950948-1G
4-Chlor-1,3-dimethyl-1H-pyrazol 95%
4-Chloro-1,3-dimethyl-1H-pyrazole
Formel: C₅H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09743376
CAS-Nummer: 15878-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR