4-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid 97%
Lieferant: Apollo Scientific
OR950952-1G
APOSOR950952-1GEA
85.7
EUR
APOSOR950952-1G
4-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid 97%
4-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid
Formel:
C₅H₅ClN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01821187 CAS-Nummer: 84547-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...