5-Ethylthiophene-2-carboxylic acid 98%

Lieferant: Apollo Scientific

OR950970-1G
APOSOR950970-1GEA 0 EUR
APOSOR950970-1G
5-Ethylthiophene-2-carboxylic acid 98%
5-Ethylthiophene-2-carboxylic acid
Formel: C₇H₈O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00130089
CAS-Nummer: 23229-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR